siRNA Details
| Gene Symbol: Scgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201561.1 | NP_963855.1 | 11 | 716 | ||
| XM_008771653.1 | XP_008769875.1 | 12 | 1122 | |||
| GenBank mRNA | AY513659.1 | - | 11 | 716 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgn| Gene Symbol: Scgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201561.1 | NP_963855.1 | 11 | 716 | ||
| XM_008771653.1 | XP_008769875.1 | 12 | 1122 | |||
| GenBank mRNA | AY513659.1 | - | 11 | 716 | ||