siRNA Details
| Gene Symbol: Akr1e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008342.1 | NP_001008343.1 | 5 | 493 | ||
| XM_008771865.1 | XP_008770087.1 | 4 | 396 | |||
| GenBank mRNA | BC086397.1 | - | 5 | 493 | 7302893 | |
| FQ212583.1 | - | 5 | 506 | |||

Akr1e2
Genomic Map