siRNA Details
| Gene Symbol: Neto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107371.1 | NP_001100841.1 | 7, 8 | 1807 | ||
| XM_006255035.2 | XP_006255097.1 | 7, 8 | 881 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Neto1| Gene Symbol: Neto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107371.1 | NP_001100841.1 | 7, 8 | 1807 | ||
| XM_006255035.2 | XP_006255097.1 | 7, 8 | 881 | |||