siRNA Details
| Gene Symbol: Tnfaip8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107387.1 | NP_001100857.1 | 3, 4 | 497 | ||
| XM_008772149.1 | XP_008770371.1 | 1, 2 | 2487 | |||
| GenBank mRNA | FQ230976.1 | - | 1, 2 | 167 | ||
| FQ235239.1 | - | 1, 2 | 167 | |||

Tnfaip8
Genomic Map