siRNA Details
| Gene Symbol: Wdr55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017932.1 | NP_001017932.1 | 4 | 484 | ||
| GenBank mRNA | BC127460.1 | - | 4 | 439 | 8367400 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr55| Gene Symbol: Wdr55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017932.1 | NP_001017932.1 | 4 | 484 | ||
| GenBank mRNA | BC127460.1 | - | 4 | 439 | 8367400 | |