siRNA Details
| Gene Symbol: Ammecr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107399.1 | NP_001100869.1 | 2, 3 | 237 | ||
| XM_006254590.2 | XP_006254652.1 | 2, 3 | 1051 | |||
| XM_006254591.2 | XP_006254653.1 | 2, 3 | 1088 | |||
| XM_006254593.2 | XP_006254655.1 | 2, 3 | 488 | |||
| XM_008772042.1 | XP_008770264.1 | 2, 3 | 1088 | |||
| GenBank mRNA | BC168199.1 | - | 2, 3 | 622 | 7113894 | |

Ammecr1l
Genomic Map