siRNA Details
| Gene Symbol: Lpcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271344.1 | NP_001258273.1 | 10 | 1034 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lpcat2| Gene Symbol: Lpcat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271344.1 | NP_001258273.1 | 10 | 1034 | ||