siRNA Details
| Gene Symbol: Acd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037193.1 | NP_001032270.1 | 7 | 615 | ||
| XM_008772496.1 | XP_008770718.1 | 7 | 669 | |||
| GenBank mRNA | BC099222.1 | - | 7 | 615 | 7388035 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acd| Gene Symbol: Acd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037193.1 | NP_001032270.1 | 7 | 615 | ||
| XM_008772496.1 | XP_008770718.1 | 7 | 669 | |||
| GenBank mRNA | BC099222.1 | - | 7 | 615 | 7388035 | |