siRNA Details
Gene Symbol: Ssx2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175597.3 | NP_783187.1 | 13, 14 | 1879 | ||
XM_006233454.2 | XP_006233516.1 | 12, 13 | 1692 | |||
XM_006233455.2 | XP_006233517.1 | 12, 13 | 1841 | |||
XM_006233458.2 | XP_006233520.1 | 9, 10 | 1822 | |||
XM_008761498.1 | XP_008759720.1 | 11, 12 | 1747 | |||
GenBank mRNA | AF532970.1 | - | 11, 12 | 1592 | ||
BC078687.1 | - | 13, 14 | 1879 | 7115887 |
