siRNA Details
| Gene Symbol: Ssx2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175597.3 | NP_783187.1 | 15 | 2190 | ||
| XM_006233454.2 | XP_006233516.1 | 14 | 2003 | |||
| XM_006233455.2 | XP_006233517.1 | 14 | 2152 | |||
| XM_006233458.2 | XP_006233520.1 | 11 | 2133 | |||
| XM_008761498.1 | XP_008759720.1 | 13 | 2058 | |||
| GenBank mRNA | AF532970.1 | - | 13 | 1903 | ||
| BC078687.1 | - | 15 | 2190 | 7115887 | ||

Ssx2ip
Genomic Map