siRNA Details
| Gene Symbol: Kif13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107462.1 | NP_001100932.1 | 4, 5 | 430 | ||
| XM_006253759.2 | XP_006253821.1 | 6, 7 | 557 | |||
| XM_006253760.2 | XP_006253822.1 | 6, 7 | 557 | |||
| XM_006253762.2 | XP_006253824.1 | 6, 7 | 557 | |||
| XM_006253763.2 | XP_006253825.1 | 6, 7 | 557 | |||
| XM_006253764.2 | XP_006253826.1 | 6, 7 | 557 | |||
| XM_006253765.2 | XP_006253827.1 | 6, 7 | 557 | |||
| XM_006253766.1 | XP_006253828.1 | 4, 5 | 424 | |||

Kif13a
Genomic Map