siRNA Details
| Gene Symbol: Vom2r23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099649.1 | NP_001093119.1 | 6, 7 | 2532 | ||
| XM_008765892.1 | XP_008764114.1 | 6 | 2438 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom2r23| Gene Symbol: Vom2r23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099649.1 | NP_001093119.1 | 6, 7 | 2532 | ||
| XM_008765892.1 | XP_008764114.1 | 6 | 2438 | |||
Set Membership: |
|