siRNA Details
| Gene Symbol: Tmem150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107476.1 | NP_001100946.1 | 3 | 470 | ||
| XM_006228267.2 | XP_006228329.1 | 4 | 880 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem150b| Gene Symbol: Tmem150b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107476.1 | NP_001100946.1 | 3 | 470 | ||
| XM_006228267.2 | XP_006228329.1 | 4 | 880 | |||