siRNA Details
| Gene Symbol: Gpr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025680.1 | NP_001020851.1 | 1 | 954 | ||
| XM_006228418.1 | XP_006228480.1 | 2 | 1545 | |||
| XM_006228419.1 | XP_006228481.1 | 2 | 2869 | |||
| GenBank mRNA | BC099196.1 | - | 1 | 954 | 7383735 | |

Gpr4
Genomic Map