siRNA Details
| Gene Symbol: Pinlyp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107488.1 | NP_001100958.1 | 5 | 579 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pinlyp| Gene Symbol: Pinlyp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107488.1 | NP_001100958.1 | 5 | 579 | ||