siRNA Details
| Gene Symbol: Acpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107510.1 | NP_001100980.1 | 5 | 507 | ||
| XM_006229051.2 | XP_006229113.1 | 7 | 855 | |||
| XM_006229052.2 | XP_006229114.1 | 8 | 776 | |||
| XM_006229053.2 | XP_006229115.1 | 7 | 891 | |||
| XM_006229054.2 | XP_006229116.1 | 7 | 927 | |||
| XM_008759366.1 | XP_008757588.1 | 6 | 668 | |||
| XM_008759367.1 | XP_008757589.1 | 7 | 891 | |||
| XM_008759368.1 | XP_008757590.1 | 7 | 888 | |||
| XM_008759369.1 | XP_008757591.1 | 6 | 1720 | |||
| GenBank mRNA | BC168932.1 | - | 7 | 932 | ||

Acpt
Genomic Map