siRNA Details
Gene Symbol: Lrrc4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271081.1 | NP_001258010.1 | 2 | 359 | ||
XM_006229055.2 | XP_006229117.1 | 2 | 411 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrrc4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271081.1 | NP_001258010.1 | 2 | 359 | ||
XM_006229055.2 | XP_006229117.1 | 2 | 411 |