siRNA Details
| Gene Symbol: Grwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012067.1 | NP_001012067.1 | 7 | 1275 | ||
| GenBank mRNA | BC083883.1 | - | 7 | 1275 | 7131596 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grwd1| Gene Symbol: Grwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012067.1 | NP_001012067.1 | 7 | 1275 | ||
| GenBank mRNA | BC083883.1 | - | 7 | 1275 | 7131596 | |