siRNA Details
| Gene Symbol: Cyfip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107517.1 | NP_001100987.1 | 2, 3 | 201 | ||
| XM_006229288.2 | XP_006229350.1 | 2, 3 | 221 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyfip1| Gene Symbol: Cyfip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107517.1 | NP_001100987.1 | 2, 3 | 201 | ||
| XM_006229288.2 | XP_006229350.1 | 2, 3 | 221 | |||