siRNA Details
| Gene Symbol: Mesdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013149.1 | NP_001013167.1 | 1 | 882 | ||
| GenBank mRNA | BC091313.1 | - | 1 | 882 | 7312243 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mesdc1| Gene Symbol: Mesdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013149.1 | NP_001013167.1 | 1 | 882 | ||
| GenBank mRNA | BC091313.1 | - | 1 | 882 | 7312243 | |