siRNA Details
| Gene Symbol: Lrtomt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139494.1 | NP_001132966.1 | 2 | 347 | ||
| GenBank mRNA | EU627093.1 | - | 2 | 347 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrtomt| Gene Symbol: Lrtomt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139494.1 | NP_001132966.1 | 2 | 347 | ||
| GenBank mRNA | EU627093.1 | - | 2 | 347 | ||