siRNA Details
| Gene Symbol: Znhit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107574.1 | NP_001101044.1 | 1 | 690 | ||
| GenBank mRNA | BC168685.1 | - | 1 | 663 | 7930608 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Znhit2| Gene Symbol: Znhit2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107574.1 | NP_001101044.1 | 1 | 690 | ||
| GenBank mRNA | BC168685.1 | - | 1 | 663 | 7930608 | |