siRNA Details
| Gene Symbol: Rcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013152.1 | NP_001013170.1 | 2 | 337 | ||
| XM_006231208.2 | XP_006231270.1 | 6 | 783 | |||
| GenBank mRNA | AY310155.1 | - | 773 | |||
| AY325146.1 | - | 773 | ||||
| BC091413.1 | - | 2 | 337 | 7326310 | ||

Rcl1
Genomic Map