siRNA Details
| Gene Symbol: Rcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013152.1 | NP_001013170.1 | 1, 2 | 240 | ||
| XM_006231208.2 | XP_006231270.1 | 5, 6 | 686 | |||
| GenBank mRNA | BC091413.1 | - | 1, 2 | 240 | 7326310 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rcl1| Gene Symbol: Rcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013152.1 | NP_001013170.1 | 1, 2 | 240 | ||
| XM_006231208.2 | XP_006231270.1 | 5, 6 | 686 | |||
| GenBank mRNA | BC091413.1 | - | 1, 2 | 240 | 7326310 | |