siRNA Details
| Gene Symbol: Ankrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107589.1 | NP_001101059.1 | 2 | 127 | ||
| GenBank mRNA | FQ216123.1 | - | 2 | 137 | ||
| FQ216298.1 | - | 2 | 137 | |||
| FQ216609.1 | - | 2 | 139 | |||
| FQ216805.1 | - | 2 | 137 | |||

Ankrd2
Genomic Map