siRNA Details
Gene Symbol: Nkx2-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107594.1 | NP_001101064.1 | 2 | 364 | ||
XM_008760427.1 | XP_008758649.1 | 3 | 1584 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nkx2-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107594.1 | NP_001101064.1 | 2 | 364 | ||
XM_008760427.1 | XP_008758649.1 | 3 | 1584 |