siRNA Details
| Gene Symbol: Ppp1r3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012072.1 | NP_001012072.1 | 2 | 842 | ||
| XM_008760370.1 | XP_008758592.1 | 2 | 1286 | |||
| GenBank mRNA | BC085894.1 | - | 2 | 842 | 7192942 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r3c| Gene Symbol: Ppp1r3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012072.1 | NP_001012072.1 | 2 | 842 | ||
| XM_008760370.1 | XP_008758592.1 | 2 | 1286 | |||
| GenBank mRNA | BC085894.1 | - | 2 | 842 | 7192942 | |