siRNA Details
| Gene Symbol: Med23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277054.1 | NP_001263983.1 | 20, 21 | 2686 | ||
| XM_008758612.1 | XP_008756834.1 | 21, 22 | 3477 | |||
| XM_008758613.1 | XP_008756835.1 | 19, 20 | 3362 | |||
| XM_008758614.1 | XP_008756836.1 | 21, 22 | 3478 | |||
| GenBank mRNA | BC090022.1 | - | 2, 3 | 348 | ||

Med23
Genomic Map