siRNA Details
Gene Symbol: Tspyl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012075.1 | NP_001012075.1 | 1 | 638 | ||
GenBank mRNA | BC082023.1 | - | 1 | 638 | 7129838 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tspyl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012075.1 | NP_001012075.1 | 1 | 638 | ||
GenBank mRNA | BC082023.1 | - | 1 | 638 | 7129838 |