siRNA Details
| Gene Symbol: Sirpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760820.1 | XP_008759042.1 | 2 | 231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sirpd| Gene Symbol: Sirpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760820.1 | XP_008759042.1 | 2 | 231 | ||