siRNA Details
| Gene Symbol: Larp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014036.1 | NP_001014058.1 | 9, 10 | 1126 | ||
| GenBank mRNA | BC081895.1 | - | 9, 10 | 1126 | 7110249 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Larp1b| Gene Symbol: Larp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014036.1 | NP_001014058.1 | 9, 10 | 1126 | ||
| GenBank mRNA | BC081895.1 | - | 9, 10 | 1126 | 7110249 | |