siRNA Details
| Gene Symbol: Cept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007699.1 | NP_001007700.1 | 5, 6 | 944 | ||
| XM_006233116.2 | XP_006233178.1 | 5, 6 | 872 | |||
| XM_006233118.2 | XP_006233180.1 | 5, 6 | 1709 | |||
| GenBank mRNA | BC079471.1 | - | 5, 6 | 944 | 7122765 | |
| FQ212036.1 | - | 5, 6 | 840 | |||

Cept1
Genomic Map