siRNA Details
| Gene Symbol: Mybphl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014042.1 | NP_001014064.1 | 7, 8 | 1173 | ||
| XM_006233150.2 | XP_006233212.1 | 7, 8 | 1539 | |||
| GenBank mRNA | BC087113.1 | - | 7, 8 | 1173 | 7193587 | |

Mybphl
Genomic Map