siRNA Details
| Gene Symbol: Cdc14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107718.2 | NP_001101188.2 | 15 | 2309 | ||
| NM_001134856.1 | NP_001128328.1 | 15 | 2235 | |||
| XM_008761460.1 | XP_008759682.1 | 16 | 1759 | |||
| GenBank mRNA | BC161876.1 | - | 15 | 2309 | 7115002 | |

Cdc14a
Genomic Map