siRNA Details
| Gene Symbol: Ccdc148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107732.1 | NP_001101202.1 | 10 | 1222 | ||
| XM_008761833.1 | XP_008760055.1 | 10 | 1308 | |||
| XM_008761834.1 | XP_008760056.1 | 10 | 1281 | |||
| XM_008761835.1 | XP_008760057.1 | 10 | 1805 | |||
| XM_008761836.1 | XP_008760058.1 | 10 | 1688 | |||
| XM_008761837.1 | XP_008760059.1 | 10 | 1308 | |||

Ccdc148
Genomic Map