siRNA Details
Gene Symbol: Slc43a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107743.1 | NP_001101213.1 | 6 | 784 | ||
XM_006234467.2 | XP_006234529.2 | 6 | 798 | |||
XM_006234468.2 | XP_006234530.2 | 6 | 710 | |||
XM_006234470.2 | XP_006234532.1 | 6 | 657 | |||
XM_008761991.1 | XP_008760213.1 | 6 | 662 |
