siRNA Details
| Gene Symbol: Aven | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107757.1 | NP_001101227.1 | 4 | 429 | ||
| XM_006234707.2 | XP_006234769.1 | 5 | 803 | |||
| XM_008762093.1 | XP_008760315.1 | 5 | 803 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aven| Gene Symbol: Aven | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107757.1 | NP_001101227.1 | 4 | 429 | ||
| XM_006234707.2 | XP_006234769.1 | 5 | 803 | |||
| XM_008762093.1 | XP_008760315.1 | 5 | 803 | |||