siRNA Details
| Gene Symbol: Adal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014047.1 | NP_001014069.1 | 5 | 699 | ||
| XM_008762166.1 | XP_008760388.1 | 10 | 1177 | |||
| XM_008762167.1 | XP_008760389.1 | 8 | 707 | |||
| XM_008762168.1 | XP_008760390.1 | 6 | 608 | |||
| XM_008762169.1 | XP_008760391.1 | 9 | 913 | |||
| GenBank mRNA | BC089959.1 | - | 5 | 699 | 7313149 | |

Adal
Genomic Map