siRNA Details
| Gene Symbol: Nanp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009409.1 | NP_001009409.1 | 2 | 692 | ||
| GenBank mRNA | BC087587.1 | - | 2 | 693 | 7319178 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nanp| Gene Symbol: Nanp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009409.1 | NP_001009409.1 | 2 | 692 | ||
| GenBank mRNA | BC087587.1 | - | 2 | 693 | 7319178 | |