siRNA Details
Gene Symbol: Prex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135718.1 | NP_001129190.1 | 10 | 1338 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
