siRNA Details
| Gene Symbol: Zfp64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012093.1 | NP_001012093.1 | 6 | 1788 | ||
| GenBank mRNA | BC085820.1 | - | 6 | 1788 | 7126205 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp64| Gene Symbol: Zfp64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012093.1 | NP_001012093.1 | 6 | 1788 | ||
| GenBank mRNA | BC085820.1 | - | 6 | 1788 | 7126205 | |