siRNA Details
| Gene Symbol: Cstf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013161.1 | NP_001013179.1 | 5 | 861 | ||
| XM_006235681.1 | XP_006235743.1 | 5 | 966 | |||
| XM_006235682.1 | XP_006235744.1 | 5 | 910 | |||
| XM_006235683.2 | XP_006235745.1 | 5 | 993 | |||
| XM_006235684.2 | XP_006235746.1 | 5 | 990 | |||
| XM_006235685.2 | XP_006235747.1 | 5 | 968 | |||
| XM_006235686.2 | XP_006235748.1 | 5 | 965 | |||
| XM_006235687.2 | XP_006235749.1 | 5 | 962 | |||
| GenBank mRNA | BC091381.1 | - | 5 | 861 | 7320917 | |

Cstf1
Genomic Map