siRNA Details
| Gene Symbol: Pbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107834.1 | NP_001101304.1 | 6, 7 | 1048 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pbx3| Gene Symbol: Pbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107834.1 | NP_001101304.1 | 6, 7 | 1048 | ||