siRNA Details
Gene Symbol: Zfp775 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107859.1 | NP_001101329.1 | 3 | 1564 | ||
XM_006236421.2 | XP_006236483.1 | 3 | 5556 | |||
XM_006236422.2 | XP_006236484.1 | 3 | 1630 | |||
XM_008762889.1 | XP_008761111.1 | 2 | 5742 |
