siRNA Details
| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_231787.7 | XP_231787.5 | 8 | 1394 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam188b| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_231787.7 | XP_231787.5 | 8 | 1394 | ||