siRNA Details
| Gene Symbol: Mcm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107873.2 | NP_001101343.2 | 7, 8 | 1284 | ||
| XM_006236863.1 | XP_006236925.1 | 7, 8 | 1394 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcm2| Gene Symbol: Mcm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107873.2 | NP_001101343.2 | 7, 8 | 1284 | ||
| XM_006236863.1 | XP_006236925.1 | 7, 8 | 1394 | |||