siRNA Details
| Gene Symbol: Mlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107889.1 | NP_001101359.1 | 2 | 337 | ||
| XM_006237363.2 | XP_006237425.1 | 3 | 327 | |||
| XM_006237364.2 | XP_006237426.1 | 3 | 327 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mlf2| Gene Symbol: Mlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107889.1 | NP_001101359.1 | 2 | 337 | ||
| XM_006237363.2 | XP_006237425.1 | 3 | 327 | |||
| XM_006237364.2 | XP_006237426.1 | 3 | 327 | |||