siRNA Details
| Gene Symbol: Tspan9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107890.1 | NP_001101360.1 | 4, 5 | 789 | ||
| XM_006237494.2 | XP_006237556.1 | 8, 9 | 920 | |||
| XM_006237495.2 | XP_006237557.1 | 7, 8 | 736 | |||
| XM_006237496.2 | XP_006237558.1 | 7, 8 | 799 | |||
| XM_006237498.2 | XP_006237560.1 | 6, 7 | 619 | |||

Tspan9
Genomic Map