siRNA Details
| Gene Symbol: Etnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107894.1 | NP_001101364.1 | 2 | 561 | ||
| XM_006237623.1 | XP_006237685.1 | 2 | 556 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etnk1| Gene Symbol: Etnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107894.1 | NP_001101364.1 | 2 | 561 | ||
| XM_006237623.1 | XP_006237685.1 | 2 | 556 | |||