siRNA Details
| Gene Symbol: Ppp1r8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107911.1 | NP_001101381.1 | 7 | 979 | ||
| GenBank mRNA | BC162025.1 | - | 7 | 978 | 9099457 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r8| Gene Symbol: Ppp1r8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107911.1 | NP_001101381.1 | 7 | 979 | ||
| GenBank mRNA | BC162025.1 | - | 7 | 978 | 9099457 | |